(E)-Buta-1,3-dien-1-ylbenzene ≥95%

Supplier: Ambeed
Danger

A1372698-1G A1372698-5G
77549-672EA 49.44 USD
77549-672 77621-922
(E)-Buta-1,3-dien-1-ylbenzene ≥95%
(E)-Buta-1,3-dien-1-ylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀
MW: 130.1864 g/mol
MDL Number: MFCD00060912
CAS Number: 16939-57-4
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR