Propargyl alcohol propoxylate 97+%

Supplier: Ambeed
Danger

Synonyms: 1-(Prop-2-yn-1-yloxy)propan-2-ol

A1372522-25G A1372522-5G
77549-668EA 40 USD
77549-668 77597-028
Propargyl alcohol propoxylate 97+%
Propargyl alcohol propoxylate

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.142 g/mol
MDL Number: MFCD09751410
CAS Number: 3973-17-9
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR