ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt
A1369194-5G
A1369194-100MG
A1369194-250MG
A1369194-1G
A1369194-25G
77549-656EA
98
USD
77549-656
77596-996
77596-998
77621-858
77867-632
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) ≥98%
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Precaución: for Research Use Only
Formula:
C₁₈H₂₄N₆O₆S₄ MW: 548.68 g/mol |
MDL Number:
MFCD00010404 CAS Number: 30931-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...