[(1,2,3-η)-2-Buten-1-yl][dicyclohexyl[3,6-dimethoxy-2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl-κC1']phosphine-κP]-Palladium(1+), 1,1,1-trifluoromethanesulfonate (1:1) ≥97%

Supplier: Ambeed
Warning

A1360917-1G A1360917-250MG A1360917-100MG A1360917-5G
77549-648EA 589 USD
77549-648 77549-650 77621-742 77867-545
[(1,2,3-η)-2-Buten-1-yl][dicyclohexyl[3,6-dimethoxy-2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl-κC1']phosphine-κP]-Palladium(1+), 1,1,1-trifluoromethanesulfonate (1:1) ≥97%
[(1,2,3-η)-2-Buten-1-yl][dicyclohexyl[3,6-dimethoxy-2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl-κC1']phosphine-κP]-Palladium(1+), 1,1,1-trifluoromethanesulfonate (1:1)

Precaución: for Research Use Only

CAS Number: 1798782-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR