(1H-Pyrazol-3-yl)methanamine ≥97%

Supplier: Ambeed
Danger

A136013-1G A136013-250MG
77549-646EA 55 USD
77549-646 77611-224
(1H-Pyrazol-3-yl)methanamine ≥97%
(1H-Pyrazol-3-yl)methanamine

Precaución: for Research Use Only

Formula: C₄H₇N₃
MDL Number: MFCD03422532
CAS Number: 37599-58-9
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR