(±)-1-Penten-3-ol ≥97%

Supplier: Ambeed
Danger

Synonyms: (±)-Ethyl vinyl carbinol, Pent-1-en-3-ol

A135718-25G A135718-5G
77549-644EA 18.54 USD
77549-644 77745-965
(±)-1-Penten-3-ol ≥97%
(±)-1-Penten-3-ol

Precaución: for Research Use Only

Formula: C₅H₁₀O
MW: 86.13 g/mol
MDL Number: MFCD00004573
CAS Number: 616-25-1
UN: 1987
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR