3-(4-Bromo-3-methyl-2-oxo-2,3-dihydro-1h-benzo[d]imidazol-1-yl)piperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A1355803-1G A1355803-5G A1355803-250MG
77549-640EA 175 USD
77549-640 77549-642 77566-662
3-(4-Bromo-3-methyl-2-oxo-2,3-dihydro-1h-benzo[d]imidazol-1-yl)piperidine-2,6-dione 98%
3-(4-Bromo-3-methyl-2-oxo-2,3-dihydro-1h-benzo[d]imidazol-1-yl)piperidine-2,6-dione

Precaución: For research use only.

Formula: C₁₃H₁₂BrN₃O₃
MDL Number: MFCD34183045
CAS Number: 2304754-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR