Fmoc-​Val-​Ala-​PAB-PNP ≥97%

Supplier: Ambeed
Warning

Synonyms: (9H-Fluoren-9-yl)methyl ((S)-3-methyl-1-(((S)-1-((4-((((4-nitrophenoxy)carbonyl)oxy)methyl)phenyl)amino)-1-oxopropan-2-yl)amino)-1-oxobutan-2-yl)carbamate, Fmoc-Val-Ala-PAB-PNP

A1353604-1G A1353604-250MG A1353604-100MG A1353604-5G
77549-636EA 540 USD
77549-636 77599-960 77596-850 77621-686
Fmoc-​Val-​Ala-​PAB-PNP ≥97%
Fmoc-​Val-​Ala-​PAB-PNP

Precaución: for Research Use Only

Formula: C₃₇H₃₆N₄O₉
MW: 680.71 g/mol
MDL Number: MFCD28557283
CAS Number: 1394238-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR