(S)-Methyl hexahydropyridazine-3-carboxylate hydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl (S)-hexahydropyridazine-3-carboxylate hydrochloride
A1352795-1G
A1352795-100MG
A1352795-250MG
77549-632EA
100
USD
77549-632
77621-670
77867-461
(S)-Methyl hexahydropyridazine-3-carboxylate hydrochloride ≥97%
(S)-Methyl hexahydropyridazine-3-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₃ClN₂O₂ MW: 180.632 g/mol |
MDL Number:
MFCD33019683 CAS Number: 380223-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...