Potassium methyl malonate 98%
Supplier: Ambeed
|
Warning
|
A135258-100G
A135258-500G
A135258-25G
A135258-1000G
A135258-5G
77549-630EA
10
USD
77549-630
77566-648
77592-556
77670-942
77883-094
Potassium methyl malonate 98%
Potassium methyl malonate
Precaución: For research use only.
Formula:
C₄H₅KO₄ |
MDL Number:
MFCD00014021 CAS Number: 38330-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...