Pentadecan-8-ol ≥95%
Supplier: Ambeed
|
Warning
|
A1337896-25G
A1337896-5G
A1337896-10G
A1337896-250MG
77549-608EA
591.22
USD
77549-608
77592-540
77596-794
77621-576
Pentadecan-8-ol ≥95%
Pentadecan-8-ol
Precaución: for Research Use Only
Formula:
C₁₅H₃₂O MW: 228.4140 g/mol |
MDL Number:
MFCD00021957 CAS Number: 1653-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...