3-Hydroxyazetidine 98%

Supplier: Ambeed
Danger

Synonyms: 3-Azetidinol, Azetidin-3-ol

A133621-1G A133621-5G A133621-250MG
77549-602EA 76.22 USD
77549-602 77566-602 77615-070
3-Hydroxyazetidine 98%
3-Hydroxyazetidine

Precaución: For research use only.

Formula: C₃H₇NO
MW: 73.09 g/mol
MDL Number: MFCD03695446
CAS Number: 45347-82-8
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR