Potassium succinate ≥98%

Supplier: Ambeed
Warning

A1330003-25G A1330003-5G
77549-596EA 129 USD
77549-596 77867-345
Potassium succinate ≥98%
Potassium succinate

Precaución: for Research Use Only

Formula: C₄H₄K₂O₄
MW: 194.2688 g/mol
MDL Number: MFCD00045954
CAS Number: 676-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR