(S)-5-((1-(3-Oxo-3-(4-(5-(trifluoromethyl)pyrimidin-2-yl)piperazin-1-yl)propoxy)propan-2-yl)amino)-4-(trifluoromethyl)pyridazin-3(2H)-one 98%

Supplier: Ambeed
Warning

Synonyms: RBN-2397

A1329221-100MG A1329221-50MG
77549-592EA 381.1 USD
77549-592 77566-596
(S)-5-((1-(3-Oxo-3-(4-(5-(trifluoromethyl)pyrimidin-2-yl)piperazin-1-yl)propoxy)propan-2-yl)amino)-4-(trifluoromethyl)pyridazin-3(2H)-one 98%
(S)-5-((1-(3-Oxo-3-(4-(5-(trifluoromethyl)pyrimidin-2-yl)piperazin-1-yl)propoxy)propan-2-yl)amino)-4-(trifluoromethyl)pyridazin-3(2H)-one

Precaución: For research use only.

Formula: C₂₀H₂₃F₆N₇O₃
MW: 523.432 g/mol
MDL Number: MFCD32701939
CAS Number: 2381037-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR