(S)-1-((S)-1-Methylpyrrolidin-2-yl)ethanol ≥97%

Supplier: Ambeed
Warning

A1329173-1G A1329173-100MG A1329173-10G A1329173-250MG A1329173-25G A1329173-5G
77549-590EA 109 USD
77549-590 77621-558 77867-327 77867-328 77867-329 77867-330
(S)-1-((S)-1-Methylpyrrolidin-2-yl)ethanol ≥97%
(S)-1-((S)-1-Methylpyrrolidin-2-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MDL Number: MFCD32263764
CAS Number: 1803485-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR