(SP-4-1)-[1,3-Bis[2,6-bis(1-propylbutyl)phenyl]-4,5-dichloro-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(3-chloropyridine-κN)-Palladium 97%

Supplier: Ambeed
Warning

A1328383-1G A1328383-250MG A1328383-5G A1328383-100MG A1328383-50MG
77549-580EA 381.1 USD
77549-580 77549-582 77549-584 77566-592 77596-772
(SP-4-1)-[1,3-Bis[2,6-bis(1-propylbutyl)phenyl]-4,5-dichloro-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(3-chloropyridine-κN)-Palladium 97%
(SP-4-1)-[1,3-Bis[2,6-bis(1-propylbutyl)phenyl]-4,5-dichloro-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(3-chloropyridine-κN)-Palladium

Precaución: For research use only.

Formula: C₄₈H₇₀Cl₅N₃Pd
MW: 972.7745 g/mol
CAS Number: 1814936-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR