4-Bromobenzylamine ≥98% (by GC)

Supplier: Ambeed
Danger

A1328373-25G A1328373-5G
77549-576EA 29.87 USD
77549-576 77549-578
4-Bromobenzylamine ≥98% (by GC)
4-Bromobenzylamine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD00047931
CAS Number: 3959-07-7
UN: 3259
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR