Doxorubicin hydrochloride 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Doxorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-10-(((2R,4S,5S,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride
A130952-1G
A130952-250MG
A130952-100MG
77549-554EA
189
USD
77549-554
77549-556
77592-512
Doxorubicin hydrochloride 99+%
Doxorubicin hydrochloride
Precaución: for Research Use Only
Formula:
C₂₇H₂₉NO₁₁·HCl |
MDL Number:
MFCD00077757 CAS Number: 25316-40-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...