Fmoc-(S)-2-(7-octenyl)alanine ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methyldec-9-enoic acid
A130771-5G
A130771-100MG
77549-550EA
963.05
USD
77549-550
77683-163
Fmoc-(S)-2-(7-octenyl)alanine ≥98%
Fmoc-(S)-2-(7-octenyl)alanine
Precaución: For research use only.
Formula:
C₂₆H₃₁NO₄ MW: 421.529 g/mol |
MDL Number:
MFCD12925760 CAS Number: 288617-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...