Fluorene 97%

Supplier: Ambeed
Danger

Synonyms: 9H-Fluorene, Diphenylenmethane

A130649-25G A130649-100G A130649-500G
77549-548EA 14.42 USD
77549-548 77566-538 77611-156
Fluorene 97%
Fluorene

Precaución: For research use only.

Formula: C₁₃H₁₀
MW: 166.22 g/mol
MDL Number: MFCD00001111
CAS Number: 86-73-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR