tert-Butyl 4-methylpyrazole-1-carboxylate ≥97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-methyl-1H-pyrazole-1-carboxylate

A130327-1G A130327-5G
77549-542EA 104.03 USD
77549-542 77745-937
tert-Butyl 4-methylpyrazole-1-carboxylate ≥97%
tert-Butyl 4-methylpyrazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
MDL Number: MFCD24466467
CAS Number: 121669-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR