2-(5-Bromopyrimidin-2-yl)acetonitrile 98%

Supplier: Ambeed
Warning

A129868-250MG A129868-1G A129868-5G
77549-540EA 132 USD
77549-540 77566-522 77871-600
2-(5-Bromopyrimidin-2-yl)acetonitrile 98%
2-(5-Bromopyrimidin-2-yl)acetonitrile

Precaución: For research use only.

Formula: C₆H₄BrN₃
MDL Number: MFCD11111850
CAS Number: 831203-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR