2,5-Dihydrofuran-3-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A1281411-1G A1281411-5G
77549-524EA 134.93 USD
77549-524 77549-526
2,5-Dihydrofuran-3-carboxylic acid ≥97%
2,5-Dihydrofuran-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆O₃
MW: 114.0993 g/mol
MDL Number: MFCD19231792
CAS Number: 1002728-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR