Palladium, chloro[(9,9-dimethyl-9H-xanthene-4,5-diyl)bis[diphenylphosphine-κP]](η3-2-propenyl)- 97%

Supplier: Ambeed
Warning

A1275547-1G A1275547-5G
77549-518EA 72.1 USD
77549-518 77566-474
Palladium, chloro[(9,9-dimethyl-9H-xanthene-4,5-diyl)bis[diphenylphosphine-κP]](η3-2-propenyl)- 97%
Palladium, chloro[(9,9-dimethyl-9H-xanthene-4,5-diyl)bis[diphenylphosphine-κP]](η3-2-propenyl)-

Precaución: For research use only.

Formula: C₄₂H₃₇ClOP₂Pd
MW: 761.560 g/mol
CAS Number: 879689-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR