2,6-Bis(benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%

Supplier: Ambeed
Warning

A1273245-10G A1273245-1G A1273245-25G A1273245-100G A1273245-5G
77549-512EA 37 USD
77549-512 77549-514 77549-516 77566-470 77592-474
2,6-Bis(benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%
2,6-Bis(benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: For research use only.

Formula: C₂₅H₂₈BNO₄
MDL Number: MFCD34182898
CAS Number: 2152673-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR