3-Bromo-4-oxo-piperidine-1-carboxylic acid benzyl ester ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Benzyl 3-bromo-4-oxopiperidine-1-carboxylate
A126518-5G
A126518-1G
77549-506EA
396.55
USD
77549-506
77611-088
3-Bromo-4-oxo-piperidine-1-carboxylic acid benzyl ester ≥98%
3-Bromo-4-oxo-piperidine-1-carboxylic acid benzyl ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₄BrNO₃ MW: 312.159 g/mol |
MDL Number:
MFCD09953304 CAS Number: 174184-13-5 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...