(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methylpent-4-enoic acid ≥97%

Supplier: Ambeed
Warning

A125933-5G A125933-100MG A125933-1G A125933-250MG
77549-498EA 795.16 USD
77549-498 77611-080 77611-082 77683-122
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methylpent-4-enoic acid ≥97%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methylpent-4-enoic acid

Precaución: For research use only.

Formula: C₂₁H₂₁NO₄
MW: 351.4 g/mol
MDL Number: MFCD08063323
CAS Number: 288617-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR