3-Bromothiophene ≥98%

Supplier: Ambeed
Danger

Synonyms: 3-Thienyl bromide

A125606-25G A125606-100G
77549-490EA 20.6 USD
77549-490 77592-438
3-Bromothiophene ≥98%
3-Bromothiophene

Precaución: for Research Use Only

Formula: C₄H₃BrS
MW: 163.04 g/mol
MDL Number: MFCD00005464
CAS Number: 872-31-1
UN: 1992
ADR: 3,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR