3-Hydroxy-1-(4-methoxybenzyl)piperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A1255505-1G A1255505-5G A1255505-250MG A1255505-25G A1255505-10G A1255505-100G
77549-486EA 68 USD
77549-486 77549-488 77566-418 77566-420 77571-526 77867-048
3-Hydroxy-1-(4-methoxybenzyl)piperidine-2,6-dione 98%
3-Hydroxy-1-(4-methoxybenzyl)piperidine-2,6-dione

Precaución: For research use only.

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD34181695
CAS Number: 2357109-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR