5-Bromo-4-fluoropyridin-2-ol ≥98%

Supplier: Ambeed
Warning

A1234510-1G A1234510-250MG A1234510-100MG
77549-456EA 65 USD
77549-456 77621-190 77866-936
5-Bromo-4-fluoropyridin-2-ol ≥98%
5-Bromo-4-fluoropyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₃BrFNO
MDL Number: MFCD28739491
CAS Number: 1805590-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR