(3-Fluoro-1-bicyclo[1.1.1]pentanyl)methanamine hydrochloride ≥98%

Supplier: Ambeed
Warning

A1233311-250MG A1233311-100MG
77549-454EA 264.71 USD
77549-454 77621-180
(3-Fluoro-1-bicyclo[1.1.1]pentanyl)methanamine hydrochloride ≥98%
(3-Fluoro-1-bicyclo[1.1.1]pentanyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁ClFN
MW: 151.6096 g/mol
MDL Number: MFCD29767888
CAS Number: 2108917-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR