Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate 98+%

Supplier: Ambeed
Danger

A1233079-1G A1233079-5G A1233079-100MG A1233079-250MG
77549-452EA 284 USD
77549-452 77566-354 77596-502 77596-504
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate 98+%
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate

Precaución: For research use only.

Formula: C₁₄H₈BF₄Rh
MDL Number: MFCD00671775
CAS Number: 36620-11-8
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR