5,5-Difluoro-1,3-piperidinedicarboxylic acid tert-butyl ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butoxycarbonyl)-5,5-difluoropiperidine-3-carboxylic acid

A122532-1G A122532-100MG A122532-250MG A122532-5G
77549-440EA 216 USD
77549-440 77889-846 77889-847 77889-848
5,5-Difluoro-1,3-piperidinedicarboxylic acid tert-butyl ester ≥98%
5,5-Difluoro-1,3-piperidinedicarboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇F₂NO₄
MDL Number: MFCD18433630
CAS Number: 1255666-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR