[(1,2,3-η)-2-Buten-1-yl]chloro[dicyclohexyl[2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine]palladium ≥98%

Supplier: Ambeed
Danger

A1218351-10G A1218351-1G A1218351-5G
77549-428EA 1288 USD
77549-428 77549-430 77549-432
[(1,2,3-η)-2-Buten-1-yl]chloro[dicyclohexyl[2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine]palladium ≥98%
[(1,2,3-η)-2-Buten-1-yl]chloro[dicyclohexyl[2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine]palladium

Precaución: for Research Use Only

Formula: C₃₇H₅₄ClPPd
CAS Number: 1798782-02-1
UN: 3175
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR