6-(Diphenylphosphino)-2,2'-bipyridine ≥98%

Supplier: Ambeed
Warning

A119622-5G A119622-100MG A119622-1G A119622-250MG
77549-400EA 1253 USD
77549-400 77745-879 77745-880 77745-881
6-(Diphenylphosphino)-2,2'-bipyridine ≥98%
6-(Diphenylphosphino)-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₂₂H₁₇N₂P
CAS Number: 152194-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR