2,4-Dihydroxybenzene-1,3,5-tricarbaldehyde ≥98%

Supplier: Ambeed
Warning

A1195795-1G A1195795-5G
77549-398EA 60.77 USD
77549-398 77596-234
2,4-Dihydroxybenzene-1,3,5-tricarbaldehyde ≥98%
2,4-Dihydroxybenzene-1,3,5-tricarbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆O₅
MW: 194.1409 g/mol
MDL Number: MFCD24679934
CAS Number: 58343-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR