(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(tetrahydro-2H-pyran-4-yl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Ala(tetrahydropyran-4-yl)-OH

A1195538-1G A1195538-5G
77549-396EA 432.6 USD
77549-396 77566-210
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(tetrahydro-2H-pyran-4-yl)propanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(tetrahydro-2H-pyran-4-yl)propanoic acid

Precaución: For research use only.

Formula: C₂₃H₂₅NO₅
MW: 395.4483 g/mol
MDL Number: MFCD18252726
CAS Number: 368866-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR