Ethyl 4-oxotetrahydro-2H-pyran-2-carboxylate ≥95%

Supplier: Ambeed
Warning

A119417-5G A119417-1G
77549-394EA 618 USD
77549-394 77611-004
Ethyl 4-oxotetrahydro-2H-pyran-2-carboxylate ≥95%
Ethyl 4-oxotetrahydro-2H-pyran-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.17 g/mol
MDL Number: MFCD20922879
CAS Number: 287193-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR