(S)-1-(2-Aminoacetyl)-4,4-difluoropyrrolidine-2-carbonitrile hydrochloride 95%

Supplier: Ambeed
Warning

A1191547-1G A1191547-250MG A1191547-5G
77549-392EA 216.3 USD
77549-392 77566-200 77620-708
(S)-1-(2-Aminoacetyl)-4,4-difluoropyrrolidine-2-carbonitrile hydrochloride 95%
(S)-1-(2-Aminoacetyl)-4,4-difluoropyrrolidine-2-carbonitrile hydrochloride

Precaución: For research use only.

Formula: C₇H₁₀ClF₂N₃O
MW: 225.6236 g/mol
MDL Number: MFCD28400675
CAS Number: 1448440-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR