(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetamido)methyl acetate ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Gly-NH-CH2-acetyloxy

A1189995-5G A1189995-1G A1189995-10G A1189995-100MG A1189995-250MG A1189995-25G
77549-390EA 271 USD
77549-390 77549-388 77596-210 77620-698 77620-700 77620-702
(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetamido)methyl acetate ≥95%
(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetamido)methyl acetate

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₅
CAS Number: 1599440-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR