6-Bromo-2,3-dihydrobenzofuran ≥95%

Supplier: Ambeed
Warning

A118810-1G A118810-250MG
77549-378EA 120 USD
77549-378 77882-969
6-Bromo-2,3-dihydrobenzofuran ≥95%
6-Bromo-2,3-dihydrobenzofuran

Precaución: for Research Use Only

Formula: C₈H₇BrO
MDL Number: MFCD08704305
CAS Number: 189035-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR