Tris(pentafluorophenyl)borane ≥97%

Supplier: Ambeed
Warning

Synonyms: Tris(perfluorophenyl)borane

A118336-25G A118336-100G
77549-376EA 219 USD
77549-376 77871-510
Tris(pentafluorophenyl)borane ≥97%
Tris(pentafluorophenyl)borane

Precaución: for Research Use Only

Formula: C₁₈BF₁₅
MDL Number: MFCD00269813
CAS Number: 1109-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR