(9Z,12Z)-Octadeca-9,12-dienal 98%

Supplier: Ambeed
Warning

A1182265-5G A1182265-100MG A1182265-1G A1182265-250MG
77549-372EA 986.74 USD
77549-372 77566-164 77571-086 77571-088
(9Z,12Z)-Octadeca-9,12-dienal 98%
(9Z,12Z)-Octadeca-9,12-dienal

Precaución: For research use only.

Formula: C₁₈H₃₂O
MW: 264.446 g/mol
MDL Number: MFCD32706447
CAS Number: 2541-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR