1-((1S,3R,4R,7S)-7-Hydroxy-1-(hydroxymethyl)-2,5-dioxabicyclo[2.2.1]heptan-3-yl)pyrimidine-2,4(1H,3H)-dione ≥98%

Supplier: Ambeed
Warning

Synonyms: 2’-O,4’-C-Methyleneuridine

A1176749-250MG A1176749-100MG
77549-362EA 255.44 USD
77549-362 77620-522
1-((1S,3R,4R,7S)-7-Hydroxy-1-(hydroxymethyl)-2,5-dioxabicyclo[2.2.1]heptan-3-yl)pyrimidine-2,4(1H,3H)-dione ≥98%
1-((1S,3R,4R,7S)-7-Hydroxy-1-(hydroxymethyl)-2,5-dioxabicyclo[2.2.1]heptan-3-yl)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₆
MW: 256.2121 g/mol
MDL Number: MFCD12911820
CAS Number: 200435-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR