2,1,3-Benzothiadiazole-4-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: Benzo[c][1,2,5]thiadiazole-4-sulphonyl chloride

A117601-1G A117601-5G
77549-360EA 36 USD
77549-360 77685-039
2,1,3-Benzothiadiazole-4-sulfonyl chloride ≥98%
2,1,3-Benzothiadiazole-4-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃ClN₂O₂S₂
MDL Number: MFCD00068049
CAS Number: 73713-79-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR