tert-Butyldimethyl(pent-4-yn-1-yloxy)silane 98%

Supplier: Ambeed
Danger

Synonyms: 5-(tert-Butyldimethylsilyloxy)-1-pentyne

A1175600-5G A1175600-1G A1175600-250MG
77549-358EA 160.68 USD
77549-358 77566-144 77596-034
tert-Butyldimethyl(pent-4-yn-1-yloxy)silane 98%
tert-Butyldimethyl(pent-4-yn-1-yloxy)silane

Precaución: For research use only.

Formula: C₁₁H₂₂OSi
MW: 198.3773 g/mol
MDL Number: MFCD28053541
CAS Number: 61362-77-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR