(SP-4-1)-[1,3-BIs[2,6-bis(1-ethylpropyl)phenyl]-4,5-dichloro-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(2-methylpyridine)palladium 98%

Supplier: Ambeed
Warning

A1167524-1G A1167524-250MG A1167524-5G A1167524-100MG A1167524-50MG
77549-328EA 1114.46 USD
77549-328 77566-126 77571-014 77595-992 77620-384
(SP-4-1)-[1,3-BIs[2,6-bis(1-ethylpropyl)phenyl]-4,5-dichloro-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(2-methylpyridine)palladium 98%
(SP-4-1)-[1,3-BIs[2,6-bis(1-ethylpropyl)phenyl]-4,5-dichloro-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(2-methylpyridine)palladium

Precaución: For research use only.

Formula: C₄₁H₅₇Cl₄N₃Pd
MW: 840.143 g/mol
CAS Number: 1612891-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR