1-Mesityl-1H-imidazole ≥97%

Supplier: Ambeed
Warning

A1159362-1G A1159362-250MG
77549-306EA 360.5 USD
77549-306 77620-294
1-Mesityl-1H-imidazole ≥97%
1-Mesityl-1H-imidazole

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂
MW: 186.25 g/mol
MDL Number: MFCD18450016
CAS Number: 25364-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR