5-(tert-Butyl)-[1,1'-biphenyl]-2-amine ≥97%

Supplier: Ambeed
Warning

A1159120-25G A1159120-1G
77549-302EA 358.44 USD
77549-302 77620-290
5-(tert-Butyl)-[1,1'-biphenyl]-2-amine ≥97%
5-(tert-Butyl)-[1,1'-biphenyl]-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₉N
MW: 225.3288 g/mol
CAS Number: 1410783-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR