2'-Deoxythymidine-5'-triphosphate trisodium salt (dTTP-Na3) ≥99%

Supplier: Ambeed
Warning

Synonyms: Thymidine-5'-triphosphoric acid trisodium salt, dTTP-Na3, Thymidine 5'-(trisodium hydrogen triphosphate), Deoxythymidine-5'-triphosphate trisodium

A1158647-100MG A1158647-250MG
77549-296EA 149 USD
77549-296 77570-968
2'-Deoxythymidine-5'-triphosphate trisodium salt (dTTP-Na3) ≥99%
2'-Deoxythymidine-5'-triphosphate trisodium salt (dTTP-Na3)

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₁₄P₃·3Na
MDL Number: MFCD00080338
CAS Number: 27821-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR